Learn More
Proteintech IHCeasy DRAP1 Ready-To-Use IHC Kit
Shop All Proteintech Products
Click to view available options
Quantity:
50 Tests
Unit Size:
50 tests
Description
KHC1739 is a ready-to-use IHC kit for staining of DRAP1. The kit provides all reagents, from antigen retrieval to cover slip mounting, that require little to no diluting or handling prior to use. Simply apply the reagents to your sample slide according to the protocol and you're steps away from obtaining high-quality IHC data.
Specifications
Specifications
| Quantity | 50 Tests |
| Form | Liquid |