missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech IHCeasy CD13 Ready-To-Use IHC Kit
Shop All Proteintech Products
Click to view available options
Unit Size:
1 set
Description
- This kit provides all reagents, from antigen retrieval to cover slip mounting, that require little to no diluting or handling prior to use.
- Simply apply the reagents to your sample slide according to the protocol and you're steps away from obtaining high-quality IHC data.
Specifications
Specifications
| Target Species | Human, Rat, Mouse |
| Sample Type | FFPE Tissue |
| Content And Storage | Store at 2°C to 8°C |
| Packaging Quantity | 50 Tests |
| Sufficient For | 50 Reactions |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction