Learn More
Proteintech IHCeasy ARHGEF5 Ready-To-Use IHC Kit
Product Code. 18026415 Shop All Proteintech Products
Click to view available options
Quantity:
50 Tests
Unit Size:
50 tests
Product Code. 18026415
Brand: Proteintech KHC169050T
KHC1690 is a ready-to-use IHC kit for staining of ARHGEF5.
KHC1690 is a ready-to-use IHC kit for staining of ARHGEF5. The kit provides all reagents, from antigen retrieval to cover slip mounting, that require little to no diluting or handling prior to use. Simply apply the reagents to your sample slide according to the protocol and you're steps away from obtaining high-quality IHC data.
Specifications
Quantity 50 Tests
Form Liquid