missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ IHC /ICC Blocking Buffer - Low Protein

Product Code. 15546276
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15546276 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15546276 Supplier Invitrogen™ Supplier No. 00495354

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Designed for use in immunohistochemistry and immunocytochemistry protocols which require blocking of non-specific binding sites. IHC /ICC Blocking Buffer, Low Protein is recommended for tissues and antibodies with average levels of background staining.

  • Can be used both in the blocking step and as a diluent for unconjugated antibodies and organic dye-conjugated antibodies during the antibody incubation step
  • Supplied as a 1X stock solution containing sodium azide
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Blocking Buffer
For Use With (Application) Immunocytochemistry (ICC)
Physical Form Liquid
Product Line eBioscience
Quantity 50 mL

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.