missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ IHC/ICC Blocking Buffer - High Protein

Product Code. 15536276
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15536276 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15536276 Supplier Invitrogen™ Supplier No. 00495254
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Designed for use in immunohistochemistry and immunocytochemistry protocols which require blocking of non-specific binding sites. IHC/ICC Blocking Buffer, High Protein is recommended for tissues and antibodies with high background staining, nuclear antigens, and FFPE tissue.

  • Can be used both in the blocking step and as a diluent for primary and secondary antibodies during antibody incubation steps
  • Compatible with organic dye-conjugated antibodies as well as unconjugated antibodies that may be used in two and three step staining protocols
  • Supplied as a 1X stock solution
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Blocking Buffer
For Use With (Application) Immunocytochemistry (ICC)
Physical Form Liquid
Product Line eBioscience
Quantity 50 mL

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.