missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ IHC Antigen Retrieval Solution - High pH (10X)

Product Code. 15576276
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15576276 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15576276 Supplier Invitrogen™ Supplier No. 00495658
  • ONLINE EXCLUSIVE PRICE
    £233.00
    £204.65 / 500mL
    Save £28.35
    12% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed for use during the heat induced epitope retrieval (HIER) step prior to immunohistochemistry on formalin-fixed paraffin embedded tissue sections. Use in combination with heat has been shown to restore the antigenicity of proteins modified during the formalin fixation of tissue.

  • Supplied as a 10x stock solution
  • Dilution of this buffer to 1x using reagent-grade water results in a 10 mM Tris, 1 mM EDTA solution, pH 9.0
TRUSTED_SUSTAINABILITY

Specifications

Product Type IHC Antigen Retrieval Solution
Quantity 500 mL
For Use With (Application) Immunohistochemistry (IHC)
Product Line eBioscience
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.