Learn More
IgY++ (IgG)(H+L) Donkey anti-Chicken, HRP, Secondary Antibody, Proteintech
Donkey anti-Chicken Affinipure Secondary Antibody
Brand:  Proteintech SA00001-14-200UL
 View more versions of this product
Product Code. 17640297
  • £40.00 / 200µL
Specifications
IgY++ (IgG)(H+L)
HRP
Donkey
200 μL
Chicken
Liquid
ELISA, Western Blot
0.01M Sodium phosphate, 0.25M NaCl, 50% glycerol, pH7.6, with 3 mg/mL BSA
The antibody was purified from antisera by immunoaffinity chromatography using antigens coupled to agarose beads.
Secondary
Store at -20°C. Stable for one year after shipment.