missing translation for 'onlineSavingsMsg'
Learn More

IgY Antibody (7C2), Novus Biologicals™

Product Code. 18370093 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18370093 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18370093 Supplier Novus Biologicals Supplier No. NBP311194

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IgY Monoclonal specifically detects IgY in Duck, Chicken samples. It is validated for Western Blot, Immunoprecipitation, Immunoassay.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgY
Applications Western Blot, Immunoprecipitation, Immunoassay
Classification Monoclonal
Clone 7C2
Concentration 0.1 mg/ml
Conjugate Unconjugated
Dilution Western Blot, Immunoprecipitation, Immunoassay
Formulation 0.2 um filtered solution in PBS, 0.1% BSA
Host Species Mouse
Immunogen The monoclonal antibody recognizes IgY.
Purification Method Protein G purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Duck, Chicken
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG1
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.