missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGSF9 Antibody (993107) [Alexa Fluor™ 532], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB9140X
This item is not returnable.
View return policy
Description
IGSF9 Monoclonal antibody specifically detects IGSF9 in Mouse samples. It is validated for Flow Cytometry, CyTOF
Specifications
| IGSF9 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 57549 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry, CyTOF | |
| 993107 | |
| Flow Cytometry, CyTOF-ready | |
| FP18798, immunoglobulin superfamily member 9A, immunoglobulin superfamily, member 9, KIAA1355 | |
| Mouse myeloma cell line NS0-derived recombinant mouse IGSF9, Arg21-Leu730, Accession # Q05BQ1 | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction