missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGSF8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14387-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member the EWI subfamily of the immunoglobulin protein superfamily. Members of this family contain a single transmembrane domain, an EWI (Glu-Trp-Ile)-motif and a variable number of immunoglobulin domains. This protein interacts with the tetraspanins CD81 and CD9 and may regulate their role in certain cellular functions including cell migration and viral infection. The encoded protein may also function as a tumor suppressor by inhibiting the proliferation of certain cancers. Alternate splicing results in multiple transcript variants that encode the same protein.Specifications
| IGSF8 | |
| Polyclonal | |
| Unconjugated | |
| IGSF8 | |
| CD316, CD81 partner 3, CD81P3, EWI 2, EWI2, IGSF8, KCT 4, KCT4, LIR D1, PGRL | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 140559, 304979, 93185 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.16 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8R366, Q969P0 | |
| IGSF8 | |
| IGSF8 Fusion Protein Ag5846 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction