Learn More
Invitrogen™ IGSF8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA532874
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Bovine, Bat, Elephant, Panda, Rabbit, Pig (100%); Mouse, Rat (93%). Immunohistochemistry with formalin-fixed paraffin-embedded tissues requires pretreatment using HIER with citrate buffer.
This gene encodes a member the EWI subfamily of the immunoglobulin protein superfamily. Members of this family contain a single transmembrane domain, an EWI (Glu-Trp-Ile)-motif and a variable number of immunoglobulin domains. This protein interacts with the tetraspanins CD81 and CD9 and may regulate their role in certain cellular functions including cell migration and viral infection. The encoded protein may also function as a tumor suppressor by inhibiting the proliferation of certain cancers. Alternate splicing results in multiple transcript variants that encode the same protein.
Specifications
| IGSF8 | |
| Polyclonal | |
| Unconjugated | |
| IGSF8 | |
| AA033172; AU041109; CD316; CD81 partner 3; CD81P3; ESTM34; EWI2; EWI-2; Glu-Trp-Ile EWI motif-containing protein 2; IGSF8; Immunoglobulin superfamily member 8; immunoglobulin superfamily, member 8; Kct4; KCT-4; Keratinocyte-associated transmembrane protein 4; keratinocytes-associated transmembrane protein 4; LIR-D1; PG regulatory-like protein; PGRL; Prostaglandin regulatory-like protein; R75384 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100154024, 100338663, 519266, 93185 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q969P0 | |
| IGSF8 | |
| Synthetic 15 amino acid peptide from internal region of human IGSF8. | |
| 50 μg | |
| Primary | |
| Human, Rabbit, Monkey, Bovine, Pig, Bat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.