missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGSF8 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA5140182
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: RLVAQLDTEG VGSLGPGYEG RHIAMEKVAS RTYRLRLEAA RPGDAGTYRC LAKAYVRGSG TRLREAASAR SRPLPVHVRE
This gene encodes a member the EWI subfamily of the immunoglobulin protein superfamily. Members of this family contain a single transmembrane domain, an EWI (Glu-Trp-Ile)-motif and a variable number of immunoglobulin domains. This protein interacts with the tetraspanins CD81 and CD9 and may regulate their role in certain cellular functions including cell migration and viral infection. The encoded protein may also function as a tumor suppressor by inhibiting the proliferation of certain cancers. Alternate splicing results in multiple transcript variants that encode the same protein.Specifications
IGSF8 | |
Polyclonal | |
Unconjugated | |
IGSF8 | |
AA033172; AU041109; CD316; CD81 partner 3; CD81P3; ESTM34; EWI2; EWI-2; Glu-Trp-Ile EWI motif-containing protein 2; IGSF8; Immunoglobulin superfamily member 8; immunoglobulin superfamily, member 8; Kct4; KCT-4; Keratinocyte-associated transmembrane protein 4; keratinocytes-associated transmembrane protein 4; LIR-D1; PG regulatory-like protein; PGRL; Prostaglandin regulatory-like protein; R75384 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
93185 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 μg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q969P0 | |
IGSF8 | |
Recombinant protein corresponding to Human IGSF8 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |