missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IGSF8 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA547450
This item is not returnable.
View return policy
Description
In direct ELISAs, approximately 15% cross-reactivity with recombinant human IGSF8 is observed and less than 1% cross-reactivity with recombinant mouse IGSF4 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
This gene encodes a member the EWI subfamily of the immunoglobulin protein superfamily. Members of this family contain a single transmembrane domain, an EWI (Glu-Trp-Ile)-motif and a variable number of immunoglobulin domains. This protein interacts with the tetraspanins CD81 and CD9 and may regulate their role in certain cellular functions including cell migration and viral infection. The encoded protein may also function as a tumor suppressor by inhibiting the proliferation of certain cancers. Alternate splicing results in multiple transcript variants that encode the same protein.
Specifications
| IGSF8 | |
| Polyclonal | |
| Unconjugated | |
| IGSF8 | |
| AA033172; AU041109; CD316; CD81 partner 3; CD81P3; ESTM34; EWI2; EWI-2; Glu-Trp-Ile EWI motif-containing protein 2; IGSF8; Immunoglobulin superfamily member 8; immunoglobulin superfamily, member 8; Kct4; KCT-4; Keratinocyte-associated transmembrane protein 4; keratinocytes-associated transmembrane protein 4; LIR-D1; PG regulatory-like protein; PGRL; Prostaglandin regulatory-like protein; R75384 | |
| Goat | |
| Antigen Affinity Chromatography | |
| RUO | |
| 140559, 304979, 93185 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q8R366, Q969P0 | |
| IGSF8 | |
| Mouse myeloma cell line NS0-derived recombinant mouse IGSF8/CD316 Ala25-Thr577. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction