missing translation for 'onlineSavingsMsg'
Learn More

IGSF8/CD316 Antibody, DyLight 594, Novus Biologicals™

Product Code. 18372614 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18372614 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18372614 Supplier Novus Biologicals Supplier No. NBP300128DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IGSF8/CD316 Polyclonal specifically detects IGSF8/CD316 in Human samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGSF8/CD316
Applications ELISA
Classification Polyclonal
Conjugate DyLight 594
Formulation 50 mM Sodium Borate
Gene Alias CD316, CD316 antigen, CD81P3LIR-D1, EWI-2, EWI2CD81 partner 3, Glu-Trp-Ile EWI motif-containing protein 2, IgSF8, immunoglobulin superfamily, member 8, KCT4, KCT-4, Keratinocytes-associated transmembrane protein 4, PGRLimmunoglobulin superfamily member 8
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant IGSF8/CD316 (Q969P0-1; Met1-Thr579).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 93185
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.