missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGSF4D/SynCAM2/CADM2 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06626PCP
This item is not returnable.
View return policy
Description
IGSF4D/SynCAM2/CADM2 Polyclonal antibody specifically detects IGSF4D/SynCAM2/CADM2 in Human samples. It is validated for ELISA
Specifications
| IGSF4D/SynCAM2/CADM2 | |
| Polyclonal | |
| ELISA | |
| cell adhesion molecule 2, IgSF4D, IGSF4DMGC138343, Immunoglobulin superfamily member 4D, immunoglobulin superfamily, member 4D, MGC104534, Necl-3, NECL3MGC138341, nectin-like 3, Nectin-like protein 3, synCAM2 | |
| Produced in rabbits immunized with purified, recombinant Human IGSF4D/SynCAM2/CADM2 (Accession#: NP_001161147.1; Gly30-Asp335) | |
| 0.1 mL | |
| Cell Biology | |
| 253559 | |
| Store at 4C in the dark. | |
| IgG |
| ELISA | |
| PerCP | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction