missing translation for 'onlineSavingsMsg'
Learn More

IGSF4D/SynCAM2/CADM2 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18341982 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18341982 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18341982 Supplier Novus Biologicals Supplier No. NBP306626AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IGSF4D/SynCAM2/CADM2 Polyclonal antibody specifically detects IGSF4D/SynCAM2/CADM2 in Human samples. It is validated for ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGSF4D/SynCAM2/CADM2
Applications ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Gene Alias cell adhesion molecule 2, IgSF4D, IGSF4DMGC138343, Immunoglobulin superfamily member 4D, immunoglobulin superfamily, member 4D, MGC104534, Necl-3, NECL3MGC138341, nectin-like 3, Nectin-like protein 3, synCAM2
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human IGSF4D/SynCAM2/CADM2 (NP_001161147.1; Gly30-Asp335).
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 253559
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.