missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGSF4C/SynCAM4 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-00164
This item is not returnable.
View return policy
Description
IGSF4C/SynCAM4 Polyclonal specifically detects IGSF4C/SynCAM4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
Specifications
| IGSF4C/SynCAM4 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| cell adhesion molecule 4, IgSF4C, IGSF4Cnectin-like 4, Immunoglobulin superfamily member 4C, immunoglobulin superfamily, member 4C, Necl-4, NECL4synCAM4, Nectin-like protein 4, SYNCAM4, TSLC1-like 2, TSLC1-like protein 2, TSLL2SynCAM4 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 199731 | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.1 - 0.3 ug/ml, Immunohistochemistry 2 - 5 ug/ml, Immunohistochemistry-Paraffin 1:10 - 1:500, Peptide ELISA Detection limit 1:16000 | |
| NP_660339.1 | |
| CADM4 | |
| Peptide with sequence C-KDERFQLEEFS corresponding to internal region according to NP_660339.1. | |
| 0.1 mg | |
| Primary | |
| Cross reacts with Human. Expected from sequence similarity: Mouse and Rat. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction