missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGSF4C/SynCAM4 Antibody (404619) [Janelia Fluor™ 525], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB4164JF525
This item is not returnable.
View return policy
Description
IGSF4C/SynCAM4 Monoclonal antibody specifically detects IGSF4C/SynCAM4 in Human samples. It is validated for Flow Cytometry,Western Blot,Immunocytochemistry,CyTOF
Specifications
| IGSF4C/SynCAM4 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| cell adhesion molecule 4, IgSF4C, IGSF4Cnectin-like 4, Immunoglobulin superfamily member 4C, immunoglobulin superfamily, member 4C, Necl-4, NECL4synCAM4, Nectin-like protein 4, SYNCAM4, TSLC1-like 2, TSLC1-like protein 2, TSLL2SynCAM4 | |
| Mouse myeloma cell line NS0-derived recombinant human IGSF4C/SynCAM4, Gln25-Tyr323, Accession # Q8NFZ8 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Western Blot, Immunocytochemistry, CyTOF | |
| 404619 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| 199731 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction