missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGSF4B/SynCAM3/CADM3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98283-50ul
This item is not returnable.
View return policy
Description
IGSF4B/SynCAM3/CADM3 Polyclonal antibody specifically detects IGSF4B/SynCAM3/CADM3 in Mouse samples. It is validated for ELISA
Specifications
| IGSF4B/SynCAM3/CADM3 | |
| Polyclonal | |
| ELISA 1:1000-1:2000 | |
| BIgR, Brain immunoglobulin receptor, cell adhesion molecule 3, IgSF4B, IGSF4BSynCAM3, Immunoglobulin superfamily member 4B, member 4B, NECL-1, NECL1TSLC1-like protein 1, Nectin-like protein 1, Synaptic cell adhesion molecule 3, synCAM3, TSLC1-like 1, TSLL1SYNCAM3 | |
| Produced in rabbits immunized with purified, recombinant Mouse IGSF4B/SynCAM3/CADM3 (Accession#: NP_444429.1; Met 1-His 328) | |
| 50 μg | |
| Neuroscience | |
| 57863 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction