missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGSF4A/SynCAM1/CADM1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£186.00 - £411.00
Specifications
| Antigen | IGSF4A/SynCAM1/CADM1 |
|---|---|
| Dilution | Western Blot 1:1000, Simple Western 1:1000, Immunocytochemistry/Immunofluorescence 1:2000 |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18244770
|
Novus Biologicals
NB300-186SS |
0.025 mL |
£186.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18748533
|
Novus Biologicals
NB300-186 |
£411.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
IGSF4A/SynCAM1/CADM1 Polyclonal specifically detects IGSF4A/SynCAM1/CADM1 in Human, Rat samples. It is validated for Western Blot, Simple Western, Immunocytochemistry/Immunofluorescence.Specifications
| IGSF4A/SynCAM1/CADM1 | |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Growth and Development, Neuronal Cell Markers, Neuroscience | |
| Whole antisera with 0.02% Sodium Azide | |
| BL2, cell adhesion molecule 1, IgSF4, IGSF4ADKFZp686F1789, IGSF4immunoglobulin superfamily, member 4D variant 2, Immunoglobulin superfamily member 4, immunoglobulin superfamily, member 4, Necl-2, NECL2MGC149785, nectin-like 2, Nectin-like protein 2, RA175, sgIGSF, Spermatogenic immunoglobulin superfamily, ST17, sTSLC-1, Synaptic cell adhesion molecule, SynCAM, synCAM1, SYNCAMMGC51880, TSLC1, TSLC-1, TSLC1/Nectin-like 2/IGSF4, Tumor suppressor in lung cancer 1SYNCAM1 | |
| CADM1 | |
| IgG | |
| Unpurified |
| Western Blot 1:1000, Simple Western 1:1000, Immunocytochemistry/Immunofluorescence 1:2000 | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human, Rat | |
| Q9BY67 | |
| 23705 | |
| A synthetic peptide corresponding to the C-terminal region of rat SynCAM/CADM1. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title