missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGSF4A/SynCAM1/CADM1 Antibody, mFluor Violet 500 SE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
IGSF4A/SynCAM1/CADM1 Polyclonal antibody specifically detects IGSF4A/SynCAM1/CADM1 in Human samples. It is validated for ELISA
Specifications
Specifications
| Purification Method | Antigen and protein A Affinity-purified |
| Research Discipline | Growth and Development, Neuronal Cell Markers, Neuroscience |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?