missing translation for 'onlineSavingsMsg'
Learn More

IGSF4A/SynCAM1/CADM1 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18388426 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18388426 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18388426 Supplier Novus Biologicals Supplier No. NBP299730AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IGSF4A/SynCAM1/CADM1 Polyclonal specifically detects IGSF4A/SynCAM1/CADM1 in Human samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGSF4A/SynCAM1/CADM1
Applications ELISA
Classification Polyclonal
Conjugate Alexa Fluor 405
Formulation 50 mM Sodium Borate
Gene Alias BL2, cell adhesion molecule 1, IgSF4, IGSF4ADKFZp686F1789, IGSF4immunoglobulin superfamily, member 4D variant 2, Immunoglobulin superfamily member 4, immunoglobulin superfamily, member 4, Necl-2, NECL2MGC149785, nectin-like 2, Nectin-like protein 2, RA175, sgIGSF, Spermatogenic immunoglobulin superfamily, ST17, sTSLC-1, Synaptic cell adhesion molecule, SynCAM, synCAM1, SYNCAMMGC51880, TSLC1, TSLC-1, TSLC1/Nectin-like 2/IGSF4, Tumor suppressor in lung cancer 1SYNCAM1
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant IGSF4A/SynCAM1/CADM1 (NP_055148.3; Met 1-His 374).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Growth and Development, Neuronal Cell Markers, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 23705
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.