missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGSF4A/SynCAM1/CADM1 Antibody (41N3D3) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
£168.00 - £387.00
Specifications
| Antigen | IGSF4A/SynCAM1/CADM1 |
|---|---|
| Clone | 41N3D3 |
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 1-5 ug/ml |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18684809
|
Novus Biologicals
NBP2-80786SS |
0.025 mg |
£168.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18292567
|
Novus Biologicals
NBP2-80786 |
0.1 mg |
£387.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IGSF4A/SynCAM1/CADM1 Monoclonal specifically detects IGSF4A/SynCAM1/CADM1 in Human samples. It is validated for Western Blot.Specifications
| IGSF4A/SynCAM1/CADM1 | |
| 0.5 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS | |
| 23705 | |
| A portion of amino acids 150-200 of human CADM1 was used as the immunogen for this antibody. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 41N3D3 | |
| Western Blot 1-5 ug/ml | |
| Unconjugated | |
| Rat | |
| Growth and Development, Neuronal Cell Markers, Neuroscience | |
| BL2, cell adhesion molecule 1, IgSF4, IGSF4ADKFZp686F1789, IGSF4immunoglobulin superfamily, member 4D variant 2, Immunoglobulin superfamily member 4, immunoglobulin superfamily, member 4, Necl-2, NECL2MGC149785, nectin-like 2, Nectin-like protein 2, RA175, sgIGSF, Spermatogenic immunoglobulin superfamily, ST17, sTSLC-1, Synaptic cell adhesion molecule, SynCAM, synCAM1, SYNCAMMGC51880, TSLC1, TSLC-1, TSLC1/Nectin-like 2/IGSF4, Tumor suppressor in lung cancer 1SYNCAM1 | |
| CADM1 | |
| IgG1 κ | |
| Protein G purified | |
| The amino acid sequence used as immunogen for this antibody is 100% homologous in human and primates. 98% in mouse, dog, rabbit, horse and Chinese hamster. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title