missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGSF3 Antibody (503621), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB4788CL1
This item is not returnable.
View return policy
Description
IGSF3 Monoclonal antibody specifically detects IGSF3 in Human samples. It is validated for Flow Cytometry,Western Blot,CyTOF
Specifications
| IGSF3 | |
| Monoclonal | |
| CoraFluor 1 | |
| EWI-3, glu-Trp-Ile EWI motif-containing protein 3, immunoglobulin superfamily, member 3, member 3, V8 | |
| Mouse myeloma cell line NS0-derived recombinant human IGSF3, Gln20-Ala1125, Accession # AAI44133 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Western Blot, CyTOF | |
| 503621 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 3321 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction