missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ IGSF2/CD101 Antibody (8D4D3), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals™ NBP3-27139-0.1ml

 View more versions of this product

Product Code. 30032757

  • £389.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

IGSF2/CD101 Monoclonal antibody specifically detects IGSF2/CD101 in Human samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IGSF2/CD101
Monoclonal
1 mg/mL
Western Blot 1:500 - 1:2000, ELISA 1:10000
CD101 antigen, CD101 molecule, Cell surface glycoprotein V7, EWI101, EWI-101, Glu-Trp-Ile EWI motif-containing protein 101, IgSF2, leukocyte surface protein, member 2
Purified recombinant fragment of human IGSF2/CD101 (AA: extra 22-168) expressed in E. Coli.
0.1 mL
Primary
Human
Purified
ELISA, Western Blot
8D4D3
Unconjugated
PBS
Mouse
Protein G purified
RUO
9398
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.