missing translation for 'onlineSavingsMsg'
Learn More

IgM Monoclonal Antibody (II/41), eFluor™ 570, eBioscience™, Invitrogen™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 41-5790-80

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15568946

  • £131.00 / 25µg
Estimated Shipment: 17-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Description: The II/41 monoclonal antibody reacts with the mu heavy chain of mouse IgM. It does not react with other classes of mouse immunoglobulin including IgD, IgG or IgA. IgM is expressed intracellularly, during early stages of B lymphopoiesis, and then on the surface of more mature B cells in the bone marrow and peripheral B cells. Fluorochrome conjugated II/41 can be used as a detection secondary for mouse IgM. Applications Reported: This II/41 antibody has been reported for use in immunohistochemical staining of frozen tissue sections, and immunocytochemistry. Applications Tested: This II/41 antibody has been tested by immunocytochemistry on fixed, differentiated OLN93 cells with O1 (Anti-Oligodendrocyte Marker) primary antibody, and by immunohistochemistry (IHC-P) on FFPE human tonsil with HI98 (Anti-CD15) as the primary antibody. This II/41 antibody can be used at less than or equal to 10 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Filter Recommendation: When using this eFluor® 570 antibody conjugate, we recommend a filter that will capture the 570 emission wavelength (for example, Excitation 545/25, 565LP, Emission 605/70). A standard Alexa Fluor® 555 or TRITC filter is acceptable. Excitation: 555 nm; Emission: 570 nm

IgM Monoclonal antibody specifically detects IgM in Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Frozen).
Specifications

Specifications

IgM
Monoclonal
0.2 mg/mL
PBS with 0.09% sodium azide; pH 7.2
P01872
Ighm
Affinity chromatography
RUO
16019
4° C, store in dark, DO NOT FREEZE!
Liquid
Immunocytochemistry, Immunohistochemistry (Frozen)
II/41
eFluor 570
Ighm
AGM1; AI326478; Igh6; Igh-6; Ighm; Igh-M; Igm; immunoglobulin heavy constant mu; MU; muH; VH
Rat
25 μg
Primary
Mouse
Antibody
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only.