missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgM Goat anti-Porcine, HRP, Polyclonal, Seconday Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP2-42699H-1000UL
This item is not returnable.
View return policy
Description
IgM Heavy Chain Polyclonal specifically detects IgM Heavy Chain in Porcine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| IgM | |
| Polyclonal | |
| HRP | |
| Phosphate Buffered Saline (PBS) containing 0.2% BSA. | |
| IGHM | |
| Pig IgM | |
| 1000 μL | |
| Secondary | |
| By immunoelectrophoresis and ELISA this antibody reacts specifically with Pig IgM. This antibody may cross react with IgM from other species. | |
| Store at 4°C. Do not freeze. |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence | |
| 1.0 mg/mL | |
| Western Blot 1:200 - 1:20000, ELISA 1:10000 - 1:100000, Immunohistochemistry 1:200 - 1:2000, Immunocytochemistry/Immunofluorescence 1:200 - 1:2000 | |
| AGM1, DKFZp686I15196, DKFZp686I15212, FLJ00385, immunoglobulin heavy constant mu, MGC104996, MGC52291, MU, VH | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3507 | |
| Pig | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction