missing translation for 'onlineSavingsMsg'
Learn More

IgM Antibody (II/41) - BSA Free, Novus Biologicals™

Rat Monoclonal Secondary Antibody

Brand:  Novus Biologicals NBP1-43303-0.025MG

122.40 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.


 View more versions of this product

Product Code. 18206955

  • £157.00 / 25µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

IgM Monoclonal specifically detects IgM in Mouse samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Mouse IgM
Monoclonal
0.5 mg/mL
Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Frozen
Constant region of heavy chain of IgM, DKFZp686I15196, DKFZp686I15212, FLJ00385, Hepatitis B virus receptor binding protein, Ig mu chain C region, IGHM, Immunoglobin heavy constant mu, Immunoglobulin heavy constant gamma 3, Immunoglobulin heavy mu chain, immunoglobulin mu, Immunoglobulin mu chain, Imunoglobulin heavy chain, MGC104996, MGC52291, mu, -region CH4 membrane bound domain, VH
Rat
Affinity Purified
RUO
Secondary
Mouse
IgG2a κ
Immunohistochemistry, Flow Cytometry, Immunohistochemistry (Frozen), ELISA
II/41
Unconjugated
Aqueous buffer, may contain carrier protein/stabilizer (proprietary). with 0.09% Sodium Azide
IGHM
The immunogen for this antibody was IgM.
0.025 mg
B Cell Development and Differentiation Markers, Immunology
3507
Store at 4C. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only