missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ iGlow™ Sample Tray Holder

Product Code. 31941218
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31941218 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31941218 Supplier Invitrogen™ Supplier No. IGA014
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Invitrogen iGlow Sample Tray Holder organizes storage of iGlow Gel Documentation System accessories. This aluminum stand features slots to accommodate the UV (IGA011), Blue (IGA012), and White (IGA013) sample trays and the iGlow UV Safety Guard (IGA015).

The Invitrogen iGlow Sample Tray Holder organizes storage of iGlow Gel Documentation System accessories. This aluminum stand features slots to accommodate the UV (IGA011), Blue (IGA012), and White (IGA013) sample trays and the iGlow UV Safety Guard (IGA015). The robust construction enables secure storage of these components in a visible and convenient location, facilitating efficient lab work and protecting valuable accessories from damage or misplacement.

Tray Holder Features and Specifications

The iGlow tray holder is made with shaped and treated aluminum. This enables consistent performance even under heavy usage in laboratory conditions.

The compact design of the tray holder enables storage of all iGlow accessories in a small space, preserving bench space while protecting components from damage or misplacement. This upright configuration also enables high visibility of the sample trays and helps ensure easy handling and storage.

TRUSTED_SUSTAINABILITY

Specifications

Product Line iGlow
For Use With (Application) DNA and Visible Gel Imaging
Product Type Gel Documentation Kit
Quantity 1 Ea.
Color White
For Use With (Equipment) iGlow Gel Documentation System
Includes Storage space for 4 imaging accessories

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.