missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IGLC2 Monoclonal Antibody (K40033_4F3), Invitrogen™
Mouse Monoclonal Antibody
£380.00 - £570.00
Specifications
| Antigen | IGLC2 |
|---|---|
| Clone | K40033_4F3 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30204933
|
Invitrogen™
MA555789 |
50 μL |
£380.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30210268
|
Invitrogen™
MA556169 |
200 μL |
£570.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: LFPPSSEELQ ANKATLVCLI SDFYPGAVTV AWKADSSPVK AGVETTTPSK QSNNKYAASS YLSLTPEQWK SHRSYSCQVT HEGSTVEKTV
IGLC2 Monoclonal antibody specifically detects IGLC2 in Human samples. It is validated for Western BlotSpecifications
| IGLC2 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| IGLC | |
| Recombinant human IGLC2 (immunogen range is 11-100). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| IGLC2 |
| K40033_4F3 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P0DOY2 | |
| 3538 | |
| IgG2a | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title