missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgK Antibody, Novus Biologicals™
Chicken Polyclonal Antibody
£288.00
Specifications
| Antigen | IgK |
|---|---|
| Dilution | Western Blot 1:100-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
IgK Polyclonal specifically detects IgK in Human samples. It is validated for Western Blot.Specifications
| IgK | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| 50802 | |
| Human IgK (Kappa) native protein. | |
| Affinity Purified | |
| Ig k |
| Western Blot 1:100-1:2000 | |
| Polyclonal | |
| Chicken | |
| Human | |
| IGK@, immunoglobulin kappa locus | |
| IGK | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title