missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IGHMBP2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17249153 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17249153 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17249153 Supplier Invitrogen™ Supplier No. PA5116653

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Samples: HepG2, A-549, Mouse brain, Mouse testis, Rat brain.

5' to 3' helicase that unwinds RNA and DNA duplices in an ATP-dependent reaction. Acts as a transcription regulator. Required for the transcriptional activation of the flounder liver-type antifreeze protein gene. Exhibits strong binding specificity to the enhancer element B of the flounder antifreeze protein gene intron. Binds to the insulin II gene RIPE3B enhancer region. May be involved in translation. DNA-binding protein specific to 5'-phosphorylated single-stranded guanine-rich sequence related to the immunoglobulin mu chain switch region. Preferentially binds to the 5'-GGGCT-3' motif. Interacts with tRNA-Tyr. Stimulates the transcription of the human neurotropic virus JCV.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGHMBP2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2.72 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene IGHMBP2
Gene Accession No. P38935, P40694, Q9EQN5
Gene Alias AEP; antifreeze enhancer-binding protein; antifreeze enhancer-binding protein ortholog; ATP-dependent helicase IGHMBP2; cardiac transcription factor 1; CATF1; CMT2S; DNA-binding protein SMUBP-2; GF-1; Glial factor 1; HCSA; HMN6; Ighmbp2; immunoglobulin mu binding protein 2; immunoglobulin mu DNA binding protein 2; immunoglobulin mu-binding protein 2; immunoglobulin S mu binding protein 2; neuromuscular degeneration; nmd; p110 subunit; RIPE3b1; sma; SMARD1; Smbp2; Smbp-2; SMUBP2; ZFAND7; zinc finger, AN1-type domain 7
Gene Symbols IGHMBP2
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20589, 29532, 3508
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.