missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IGHMBP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568960
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rat: 100%.
5' to 3' helicase that unwinds RNA and DNA duplices in an ATP-dependent reaction. Acts as a transcription regulator. Required for the transcriptional activation of the flounder liver-type antifreeze protein gene. Exhibits strong binding specificity to the enhancer element B of the flounder antifreeze protein gene intron. Binds to the insulin II gene RIPE3B enhancer region. May be involved in translation. DNA-binding protein specific to 5'-phosphorylated single-stranded guanine-rich sequence related to the immunoglobulin mu chain switch region. Preferentially binds to the 5'-GGGCT-3' motif. Interacts with tRNA-Tyr. Stimulates the transcription of the human neurotropic virus JCV.
Specifications
| IGHMBP2 | |
| Polyclonal | |
| Unconjugated | |
| IGHMBP2 | |
| AEP; antifreeze enhancer-binding protein; antifreeze enhancer-binding protein ortholog; ATP-dependent helicase IGHMBP2; cardiac transcription factor 1; CATF1; CMT2S; DNA-binding protein SMUBP-2; GF-1; Glial factor 1; HCSA; HMN6; Ighmbp2; immunoglobulin mu binding protein 2; immunoglobulin mu DNA binding protein 2; immunoglobulin mu-binding protein 2; immunoglobulin S mu binding protein 2; neuromuscular degeneration; nmd; p110 subunit; RIPE3b1; sma; SMARD1; Smbp2; Smbp-2; SMUBP2; ZFAND7; zinc finger, AN1-type domain 7 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 20589 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P40694 | |
| IGHMBP2 | |
| synthetic peptide corresponding to a region of mouse IGHMBP2 (aa 15-64). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction