missing translation for 'onlineSavingsMsg'
Learn More

IGHMBP2 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18380322 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18380322 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18380322 Supplier Novus Biologicals Supplier No. NBP306533JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IGHMBP2 Polyclonal antibody specifically detects IGHMBP2 in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGHMBP2
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias ATP-dependent helicase IGHMBP2, CATF1FLJ41171, EC 3.6.1, EC 3.6.4.12, EC 3.6.4.13, GF-1, Glial factor 1, HCSA, HMN6cardiac transcription factor 1, immunoglobulin mu binding protein 2, Immunoglobulin mu-binding protein 2, SMARD1DNA-binding protein SMUBP-2, SMBP2, SMUBP2FLJ34220
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human IGHMBP2 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neurodegeneration, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 3508
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.