missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGHG1 Rabbit anti-Human, Clone-IG1707R, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-54449-100UG
This item is not returnable.
View return policy
Description
IgG Heavy Chain Monoclonal specifically detects IgG Heavy Chain in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| IGHG1 | |
| Monoclonal | |
| 1 mg/mL | |
| Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, CyTOF-ready | |
| immunoglobulin heavy constant gamma 1 (G1m marker) | |
| Rabbit | |
| 75 kDa | |
| 100 μg | |
| Immunology | |
| 3500 | |
| Store at -20 to -80C. Avoid freeze-thaw cycles. |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF | |
| IG1707R | |
| Unconjugated | |
| PBS with No Preservative | |
| IGHG1 | |
| Purified human Ig Gamma Chain | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction