missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG4 Fc Antibody (RM217), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-62127
This item is not returnable.
View return policy
Description
IgG4 Fc Monoclonal antibody specifically detects IgG4 Fc in Human samples. It is validated for ELISA, Sandwich ELISA Capture, Sandwich ELISA Detection
Specifications
| IgG4 Fc | |
| Monoclonal | |
| 1 mg/mL | |
| ELISA, Sandwich ELISA Capture 50 - 200ng/well, Sandwich ELISA Detection 0.05 - 0.2ug/mL | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Sandwich ELISA Capture, Sandwich ELISA Detection | |
| RM217 | |
| Unconjugated | |
| 50% Glycerol/PBS, 1% BSA | |
| Peptide corresponding to the Fc region of Human IgG4 | |
| 100 μg | |
| Immunology | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction