missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG4 Antibody (RM120), Biotin, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-62126B
This item is not returnable.
View return policy
Description
IgG4 Monoclonal specifically detects IgG4 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry.
Specifications
| IgG4 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 0.5 ug/ml - 2 ug/ml, Flow Cytometry 1ug/ml - 4ug/ml, ELISA, Immunohistochemistry 1ug/mL - 10ug/mL, Immunocytochemistry/Immunofluorescence, Immunocytochemistry 1ug/mL - 10ug/mL | |
| Immunoglobulin G, Immunoglobulin G4, ImmunoglobulinG | |
| Peptide corresponding to the hinge region of Human IgG4 | |
| 50 μg | |
| This antibody reacts to the heavy chain of human IgG4, This antibody does not cross react to any other IgG subclasses (IgG1, IgG2, or IgG3), and shows no cross reactivity to IgM, IgA, IgD, IgE. This antibody does not react to monkey (Cyno or Rhesus) IgG, mouse IgG, rat IgG, or goat IgG. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry | |
| RM120 | |
| Biotin | |
| 50% Glycerol/PBS with 1% BSA with 0.09% Sodium Azide | |
| Rabbit | |
| Protein A purified | |
| Secondary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction