missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG3 Mouse, Alexa Fluor 488, Clone: MG3-35, Isotype Control, STEMCELL Technologies
Mouse Monoclonal Antibody
£187.65 - £380.70
Specifications
Antigen | IgG3 |
---|---|
Clone | MG3-35 |
Applications | Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence |
Classification | Monoclonal |
Conjugate | Alexa Fluor 488 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17138871
|
Stemcell Technologies
60073AD.1 |
25 μg |
£187.65
25µg |
Estimated Shipment: 05-07-2024
Log in to see stock. |
|||||
17128871
|
Stemcell Technologies
60073AD |
100 μg |
£380.70
100µg |
Estimated Shipment: 05-07-2024
Log in to see stock. |
|||||
Description
- The MG3-35 antibody (IgG3, kappa) is suitable for use as an isotype-matched control antibody in several applications to estimate the degree of non-specific binding by an antigen-specific antibody
- Ideally, the isotype control should have the same subclass of heavy chain (IgA, IgD, IgE, IgG, or IgM) and light chain (kappa or lambda) as the specific antibody being used
- If a conjugated antibody is used, an isotype control conjugated to the same molecule (e.g. fluorochrome) should be chosen
- The use of an appropriate isotype control helps confirm the specificity of the antigen-specific antibody and indicates non-specific binding that may result from binding to Fc receptors or other cell components
- This antibody clone has been verified for use as an isotype control antibody for assessing non-specific binding to cells in applications such as flow cytometry and immunofluorescence microscopy applications (surface and intracellular staining)
Specifications
IgG3 | |
Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
Alexa Fluor 488 | |
IgG3 κ |
MG3-35 | |
Monoclonal | |
Mouse | |
Isotype Control (Primary) |