missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG2b Rat, FITC, Clone: RTK4530, Isotype Control, STEMCELL Technologies
Rat Monoclonal Antibody
£70.20 - £279.45
Specifications
Antigen | IgG2b |
---|---|
Clone | RTK4530 |
Applications | Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence |
Classification | Monoclonal |
Conjugate | FITC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17198911
|
Stemcell Technologies
60077FI.1 |
50 μg |
£74.25
50µg |
Estimated Shipment: 05-07-2024
Log in to see stock. |
|||||
17188911
|
Stemcell Technologies
60077FI |
200 μg |
£279.45
200µg |
Estimated Shipment: 05-07-2024
Log in to see stock. |
|||||
17118921
|
Stemcell Technologies
60077FI.3 |
25 Tests |
£70.20
25 tests |
Estimated Shipment: 05-07-2024
Log in to see stock. |
|||||
17108921
|
Stemcell Technologies
60077FI.2 |
100 Tests |
£184.95
100 tests |
Estimated Shipment: 05-07-2024
Log in to see stock. |
|||||
Description
- The RTK4530 antibody (IgG2b, kappa) is suitable for use as an isotype-matched control antibody in several applications to estimate the degree of non-specific binding by an antigen-specific antibody
- Ideally, the isotype control should have the same subclass of heavy chain (IgA, IgD, IgE, IgG, or IgM) and light chain (kappa or lambda) as the specific antibody being used
- If a conjugated antibody is used, an isotype control conjugated to the same molecule (e.g. fluorochrome) should be chosen
- The use of an appropriate isotype control helps confirm the specificity of the antigen-specific antibody and indicates non-specific binding that may result from binding to Fc receptors or other cell components
- This antibody clone has been verified for use as an isotype control antibody for assessing non-specific binding to cells in applications such as flow cytometry and immunofluorescence microscopy applications (surface and intracellular staining)
Specifications
IgG2b | |
Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
FITC | |
IgG2b κ |
RTK4530 | |
Monoclonal | |
Rat | |
Isotype Control (Primary) |