missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG2a Rat, FITC, Clone: RTK2758, Isotype Control, STEMCELL Technologies
Rat Monoclonal Antibody
£79.65 - £176.85
Specifications
Antigen | IgG2a |
---|---|
Clone | RTK2758 |
Applications | Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence |
Classification | Monoclonal |
Conjugate | FITC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17188901
|
Stemcell Technologies
60076FI.1 |
50 μg |
£79.65
50µg |
Estimated Shipment: 05-07-2024
Log in to see stock. |
|||||
17178901
|
Stemcell Technologies
60076FI |
200 μg |
£176.85
200µg |
Estimated Shipment: 05-07-2024
Log in to see stock. |
|||||
Description
- The RTK2758 antibody (IgG2a, kappa) is suitable for use as an isotype-matched control antibody in several applications to estimate the degree of non-specific binding by an antigen-specific antibody
- Ideally, the isotype control should have the same subclass of heavy chain (IgA, IgD, IgE, IgG, or IgM) and light chain (kappa or lambda) as the specific antibody being used
- If a conjugated antibody is used, an isotype control conjugated to the same molecule (e.g. fluorochrome) should be chosen
- The use of an appropriate isotype control helps confirm the specificity of the antigen-specific antibody and indicates non-specific binding that may result from binding to Fc receptors or other cell components
- This antibody clone has been verified for use as an isotype control antibody for assessing non-specific binding to cells in applications such as flow cytometry and immunofluorescence microscopy applications (surface and intracellular staining)
Specifications
IgG2a | |
Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
FITC | |
IgG2a κ |
RTK2758 | |
Monoclonal | |
Rat | |
Isotype Control (Primary) |