missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat IgG2a kappa Isotype Control (eBR2a), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
Rat Isotype Control
£76.50 - £172.00
Specifications
Antigen | Rat IgG2a kappa |
---|---|
Clone | eBR2a |
Concentration | 0.2 mg/mL |
Applications | Control, Flow Cytometry |
Conjugate | PerCP-eFluor 710 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15321320
|
Affymetrix eBioscience
46-4321-80 |
25 μg |
£76.50
25µg |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||||
15331320
|
Affymetrix eBioscience
46-4321-82 |
100 μg |
£172.00
100µg |
In Stock
Log in to see stock. |
|||||
Description
Description: The monoclonal rat IgG2a kappa is useful as an isotype control immunoglobulin. Applications Reported: PerCP-eFluor® 710 Rat IgG2a Isotype Control has been reported for use in flow cytometric analysis. Applications Tested: Rat IgG2a Isotype Control has been tested by flow cytometric analysis of mouse splenocyte suspensions. It should be used at the same concentration as the experimental antibody. Excitation: 488 nm; Emission: 710 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.
Rat IgG2a is an immunoglobulin subclass that is commonly used in research and diagnostic applications. It is produced by the rat immune system and is known for its ability to activate complement and mediate antibody-dependent cell-mediated cytotoxicity (ADCC). Rat IgG2a has a molecular weight of approximately 160 kDa and is composed of two heavy chains and two light chains. It is often used as a control or reference antibody in experiments involving rat models, and is also commonly used in flow cytometry and immunohistochemistry applications.Specifications
Rat IgG2a kappa | |
0.2 mg/mL | |
PerCP-eFluor 710 | |
Rat | |
Not Applicable | |
IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2 | |
Isotype Control (Primary) | |
4° C, store in dark, DO NOT FREEZE! |
eBR2a | |
Control, Flow Cytometry | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
IgG2a κ | |
Affinity chromatography |
For Research Use Only.