missing translation for 'onlineSavingsMsg'
Learn More

IgG1 Mouse, APC, Clone: MOPC-21, Isotype Control, STEMCELL Technologies

Mouse Monoclonal Antibody

£60.75 - £162.00

Specifications

Antigen IgG1
Clone MOPC-21
Applications Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Conjugate APC
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17198831
Encompass_Preferred
View Documents
Stemcell Technologies
60070AZ.1
25 μg
£60.75
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17188831
Encompass_Preferred
View Documents
Stemcell Technologies
60070AZ
100 μg
£162.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • The MOPC-21 antibody (IgG1, kappa) is suitable for use as an isotype-matched control antibody in several applications to estimate the degree of non-specific binding by an antigen-specific antibody
  • Ideally, the isotype control should have the same subclass of heavy chain (IgA, IgD, IgE, IgG, or IgM) and light chain (kappa or lambda) as the specific antibody being used
  • If a conjugated antibody is used, an isotype control conjugated to the same molecule (e.g. fluorochrome) should be chosen
  • The use of an appropriate isotype control helps confirm the specificity of the antigen-specific antibody and indicates non-specific binding that may result from binding to Fc receptors or other cell components
  • This antibody clone has been verified for use as an isotype control antibody for assessing non-specific binding to cells in applications such as flow cytometry and immunofluorescence microscopy applications (surface and intracellular staining)
Specifications

Specifications

IgG1
Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
APC
IgG1 κ
MOPC-21
Monoclonal
Mouse
Isotype Control (Primary)
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.