missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG1 Fc Antibody (43D17), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22028PE
This item is not returnable.
View return policy
Description
IgG1 Fc Monoclonal antibody specifically detects IgG1 Fc in Human samples. It is validated for Western Blot, ELISA
Specifications
| IgG1 Fc | |
| Monoclonal | |
| PE | |
| PBS | |
| Human recombinant IgG1 Fc | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, ELISA | |
| 43D17 | |
| Western Blot, ELISA | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction