missing translation for 'onlineSavingsMsg'
Learn More

IgG1[a] Mouse anti-Mouse, BUV615, Clone: 10.9, BD Biosciences™

Mouse Monoclonal Antibody

Brand:  BD Biosciences 751624

Product Code. 17222795

  • ONLINE EXCLUSIVE PRICE
    £192.00
    £188.32 / 50µg
    Save £3.68
    1% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

IgG1[a] Monoclonal antibody specifically detects IgG1[a] in Mouse samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IgG1[a]
Monoclonal
Brilliant Ultraviolet 615
Igh-4a; Ighg1; IgG1
Not reported
RUO
Mouse
IgG2a κ
Flow Cytometry
10.9
Aqueous buffered solution containing ≤0.09% sodium azide.
Mouse
50 μg
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.