missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human IgG (Total) Uncoated ELISA Kit

Product Code. 15510947
Click to view available options
Quantity:
10 x 96 Tests
20 x 96 Tests
Unit Size:
Pack of 10
Pack of 20
This item is not returnable. View return policy

Product Code. 15510947

Brand: Invitrogen™ 885055088

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

The Human Total Immunoglobulin G (igG) Uncoated ELISA Kit contains pre-matched antibody pairs, and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of human igG. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.

Human IgG, also known as human immunoglobulin G, is an antibody that is produced by the immune system in response to antigens. It is the most abundant immunoglobulin in the blood and plays a key role in defense against pathogens. IgG works by binding to specific antigens and promoting their clearance from the body through various mechanisms, including phagocytosis and complement activation. IgG is also involved in other immune responses, such as antibody-dependent cellular cytotoxicity and neutralization of toxins. Human IgG can be used in research to study the immune response to pathogens and to develop diagnostic tests and therapeutic interventions for infectious and autoimmune diseases.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 1.6 to 100 ng/mL
Assay Sensitivity 1.6 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum
Target Species Human
For Use With (Application) ELISA
For Use With (Equipment) Colorimetric Microplate Reader
Kit Contents Capture Antibody: Pre-titrated, Purified antibody, Detection Antibody: Pre-titrated, HRP-conjugated antibody, Standard: Protein for generating standard curve and calibrating samples, 10X Coating Buffer: Buffer for plating the Capture Antibody, 5X ELISA/ELISPOT Diluent: Buffer for blocking and diluting the Detection Antibody and Enzyme, Substrate: 1X TMB Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards, 96 Well Plate: High affinity 96-well plate (included with product Cat. Nos. ending in suffixes -22, -76, -86)
Label or Dye HRP
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 10 μL, Serum 10 μL
Storage Requirements 2°C to 8°C
Synonym Immunoglobulin G
Target Human IgG
Test Time 1 hrs 45 mins
Total Assay Time 24 hrs 30 mins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.