Learn More
IgG Heavy Chain Mouse anti-Rabbit, CoraLite 488, Clone: 1C9A5, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL488-66467-100UL
Product Code. 17571586
  • £189.00 / 100µL
Description
The Heavy chain of Rabbit IgG antibody from Proteintech is a mouse monoclonal antibody to a protein of rabbit Heavy chain of Rabbit IgG. This antibody recognizes rabbit antigen. The Heavy chain of Rabbit IgG antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specifications
Specifications
IgG Heavy Chain
Monoclonal
2000 μg/mL
Mouse
Primary
IgG1
Immunofluorescence
1C9A5
CoraLite 488