missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG Heavy Chain Antibody (RM116), Biotin, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-62019B
This item is not returnable.
View return policy
Description
IgG Heavy Chain Monoclonal specifically detects IgG Heavy Chain in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry.
Specifications
| IgG Heavy Chain | |
| Monoclonal | |
| 1 mg/mL | |
| Flow Cytometry, ELISA, Immunohistochemistry 0.5ug/ml - 2ug/ml, Immunocytochemistry/Immunofluorescence 0.5ug/ml - 2ug/ml, Immunocytochemistry 0.5 - 2 ug/mL | |
| Immunoglobulin G, ImmunoglobulinG | |
| Human IgG | |
| 50 μg | |
| This antibody reacts to the Fc region of all gamma heavy chains of human immunoglobulins, including gamma 1, gamma 2, gamma 3, and gamma 4. No cross reactivity with other human heavy chains, mouse IgG, rat IgG or goat IgG | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry | |
| RM116 | |
| Biotin | |
| PBS, 50% Glycerol, 1% BSA with 0.09% Sodium Azide | |
| Rabbit | |
| Protein A purified | |
| Secondary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction