missing translation for 'onlineSavingsMsg'
Learn More

IgG Heavy Chain Antibody (IG1707R), Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18411354 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18411354 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18411354 Supplier Novus Biologicals Supplier No. NBP254449JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

IgG Heavy Chain Monoclonal antibody specifically detects IgG Heavy Chain in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG Heavy Chain
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone IG1707R
Conjugate Janelia Fluor 646
Formulation 50 mM Sodium Borate
Gene Alias Immunoglobulin G, ImmunoglobulinG
Host Species Rabbit
Immunogen Recombinant full-length human IgG Heavy Chain protein
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.