missing translation for 'onlineSavingsMsg'
Learn More

IgG (γ-Heavy Chain) Ab-1 Rabbit Polyclonal Antibody, Epredia™

IgG (γ-Heavy Chain) Ab-1, Rabbit PAb is one marker in a comprehensive ab portfolio that addresses all major disease states ensuring the ideal tests are always available.

Brand:  Epredia RB-1432-A1

Code : LD

Additional Details : Weight : 0.00190kg

 View more versions of this product

Product Code. 12612887

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

The most common feature of plasmacytomas, and certain non-Hodgkin's lymphomas is the restricted expression of a single heavy chain class. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: IgG isolated from a pool of normal human sera

Molecular Weight: 50 kDa

Positive Control: Tonsil

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

IgG (gamma heavy Chain) Ab-1
Polyclonal
Rabbit
500 μL
Immunoglobulins
Human
IgG
Immunohistochemistry (Paraffin)
Unconjugated
IgG isolated from a pool of normal human sera
IVD
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers