missing translation for 'onlineSavingsMsg'
Learn More

IgG (γ-Heavy Chain) Ab-1 Rabbit Polyclonal Antibody, Epredia™

IgG (γ-Heavy Chain) Ab-1, Rabbit PAb is one marker in a comprehensive ab portfolio that addresses all major disease states ensuring the ideal tests are always available.

Brand:  Epredia RB-1432-A0

 View more versions of this product

Product Code. 12602887

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

The most common feature of plasmacytomas, and certain non-Hodgkin's lymphomas is the restricted expression of a single heavy chain class. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: IgG isolated from a pool of normal human sera

Molecular Weight: 50 kDa

Positive Control: Tonsil

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IgG (gamma heavy Chain) Ab-1
Polyclonal
Rabbit
100 μL
Immunoglobulins
Human
IgG
Immunohistochemistry (Paraffin)
Unconjugated
IgG isolated from a pool of normal human sera
IVD
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.